4-Benzyloxy-1-butanol 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-(Benzyloxy)butan-1-ol, 1-Butanol, 4-(phenylmethoxy)-, 4-benzyloxy-butan-1-ol, 4-benzyloxybutanol

399120050 399120250
ACRO399120050EA 448 DKK
ACRO399120050 ACRO399120250
4-Benzyloxy-1-butanol 97%
4-Benzyloxy-1-butanol
Formel: C₁₁H₁₆O₂
Kogepunkt: 216…256 °C (760 mmHg)
Massefylde: 1,026 g/cm³
Flammepunkt: 109 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00155211
CAS-nummer: 4541-14-4

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5120 to 1.5140 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR