3-Hydroxyphenylacetylene 95+%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
3-Ethynylphenol
392410010
392410050
ACRO392410010EA
757
DKK
ACRO392410010
ACRO392410050
3-Hydroxyphenylacetylene 95+%
3-Hydroxyphenylacetylene
Formel:
C₈H₆O Massefylde: 1,803 g/cm³ Flammepunkt: 103 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00078347 CAS-nummer: 10401-11-3 |
Specifikationer Testresultater
Appearance | Clear yellow to orange to light brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5840 to 1.5880 (20°C, 589 nm) |
GC | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...