Benzylideneacetone 98+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

385590010 385590500 385592500
ACRO385590010EA 673 DKK
ACRO385590010 ACRO385590500 ACRO385592500
Benzylideneacetone 98+%
Benzylideneacetone
Formel: C₁₀H₁₀O
Kogepunkt: 261 °C (760 mmHg)
Flammepunkt: 116 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008779
CAS-nummer: 122-57-6

Bestil nu

Specifikationer Testresultater

Appearance Yellow Low melting solid
Infrared spectrum Conforms
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR