Bis(dichlorophosphino)methane 96%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Phosphonous dichloride,P,P'-methylenebis-, Methylenebis(dichlorophosphine), dichloro(dichlorophosphanylmethyl)phosphane

383000010 383000050
ACRO383000010EA 665 DKK
ACRO383000010 ACRO383000050
Bis(dichlorophosphino)methane 96%
Bis(dichlorophosphino)methane
Formel: CH₂Cl₄P₂
Kogepunkt: 101…105 °C (22 mmHg)
Massefylde: 1,601 g/cm³
Flammepunkt: 96 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01073767
CAS-nummer: 28240-68-8
UN nummer: 3265
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid
Refractive index 1.5920 to 1.5950 (20°C, 589 nm)
Titration Argentometric ≥95 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR