tert-Butyl trichloroacetimidate 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: tert-Butyl 2,2,2-trichloroacetimidate, t-Butyl trichioroacetimidate, t-butyl-2,2,2-trichloroacetimidate

369330050 369330250 369331000
ACRO369330050EA 428 DKK
ACRO369330050 ACRO369330250 ACRO369331000
tert-Butyl trichloroacetimidate 95%
tert-Butyl trichloroacetimidate
Formel: C₆H₁₀Cl₃NO
Kogepunkt: 64…66 °C (15 mmHg)
Smeltepunkt: 20 °C
Massefylde: 1,222 g/cm³
Flammepunkt: 55 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00077410
CAS-nummer: 98946-18-0
UN nummer: 1993
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid or low melting crystalline mass
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4570 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR