Chlortripropylsilan 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

313930050 313930250
ACRO313930050EA 570 DKK
ACRO313930050 ACRO313930250
Chlortripropylsilan 98%
Chlorotripropylsilane
Formel: C₉H₂₁ClSi
Kogepunkt: 199…200 °C (760 mmHg)
Massefylde: 0,882 g/cm³
Flammepunkt: 76 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000508
CAS-nummer: 995-25-5
UN nummer: 2987
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4420 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR