Cadmiumacetat dihydrat 98%, pure
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Cadmium di(acetate) dihydrate, Acetic acid cadmium salt dihydrate
212310010 212310050 212312500
ACRO212310010EA 1100 DKK
ACRO212310010 ACRO212310050 ACRO212312500
Cadmiumacetat dihydrat 98%, pure
Cadmiumacetat dihydrat
Formel: (H₃CCOO)₂Cd·2H₂O
Molekylvægt: 266.53 g/mol
Kogepunkt: 105…107 °C (4 mmHg)
Smeltepunkt: 256 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00150020
CAS-nummer: 5743-04-4
UN nummer: 2570
ADR: 6.1 III
Merck Index: 13,01614
Bestil nu
Specifikationer Testresultater
Appearance White Crystals
Titration Complexometric ≥97.5 %
Loss on drying 12.5 % to 14.5 %