2-Hydroxyethylsalicylat 98+%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
174100010
174102500
ACRO174100010EA
1950
DKK
ACRO174100010
ACRO174102500
2-Hydroxyethylsalicylat 98+%
2-Hydroxyethylsalicylat
Formel:
C₉H₁₀O₄ Molekylvægt: 182.18 g/mol Kogepunkt: 172 °C (12 mmHg) Smeltepunkt: 25 °C Massefylde: 1.25 g/cm³ Flammepunkt: 174 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002862 CAS-nummer: 87-28-5 |
Specifikationer Testresultater
Appearance | Colorless to almost colorless Oily liquid or fused mass |
Infrared spectrum | Conforms |
Refractive index | 1.5480 to 1.5510 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...