Sulphur trioxide trimethylamine complex 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

159350250 159351000 159355000
ACRO159350250EA 306 DKK
ACRO159350250 ACRO159351000 ACRO159355000
Sulphur trioxide trimethylamine complex 95%
Sulphur trioxide trimethylamine complex
Formel: C₃H₉NO₃S
Smeltepunkt: 232 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012421
CAS-nummer: 3162-58-1
UN nummer: 3261
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with NaOH ≥94 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.01 %
Potassium (K) ≤0.005 %
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR