9-Phenanthrencarbaldehyd
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
9-Phenanthrenealdehyde, 9-Phenanthrenecarboxaldehyde, Phenanthrene-9-carboxaldehyde
130110010
130110050
ACRO130110010EA
275
DKK
ACRO130110010
ACRO130110050
9-Phenanthrencarbaldehyd
9-Phenanthrencarbaldehyd
Formel:
C₁₅H₁₀O Smeltepunkt: 100 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001175 CAS-nummer: 4707-71-5 |
Specifikationer Testresultater
Appearance | Light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 99 to 103 °C |
HPLC | ≥96.0 % |
UV | at 260 nm A: ≤0.02 |
Titration after Ion exchange | ≥99.0 % (on dry substance) |
Membrane filtration test | (on 0.5 M soln.) Passes test |
Suitability | (HPLC gradient) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...