Nitro blue tetrazolium monohydrat 90%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride
128590010 128590050
ACRO128590010EA 1890 DKK
ACRO128590010 ACRO128590050
Nitro blue tetrazolium monohydrat 90%
Nitro blue tetrazolium monohydrat
Formel: C₄₀H₃₀Cl₂N₁₀O₆·H₂O·CH₄O
Smeltepunkt: >170 °C (dec.)
Opbevaringstemperatur: Fryser
MDL Number: MFCD00150013
CAS-nummer: 298-83-9
Bestil nu
Specifikationer Testresultater
Appearance Yellow Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (105°C)
Sulfated ash ≤0.1 %
HPLC ≥88 %