4,5-Dimethyl-o-phenylendiamin 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4,5-Dimethylbenzene-1,2-diamine, 4,5-Dimethyl-1,2-phenylenediamine, 4,5-Diamino-1,2-dimethylbenzene, 1,2-Diamino-4,5-dimethylbenzene

116610050 116610250
ACRO116610050EA 740 DKK
ACRO116610050 ACRO116610250
4,5-Dimethyl-o-phenylendiamin 98%
4,5-Dimethyl-o-phenylendiamin
Formel: C₈H₁₂N₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007729
CAS-nummer: 3171-45-7
UN nummer: 2811
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance Yellow to orange to brown Powder or solid
Infrared spectrum Conforms
GC ≥97.5 %
Calcium (Ca) ≤0.05 %
Iron (Fe) ≤0.001 %
Potassium (K) ≤0.03 %
Sodium (Na) ≤0.05 %
Strontium (Sr) ≤0.2 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 %
Oxidizing substance (as NO3) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR