Anti-MS4A1 Mouse Monoclonal Antibody [clone: L26]
Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ILM30211-C01 ILM30211-C05 ILM30211-C1 ILM30211-R10 ILM30211-R25 S/ILM30211-R2
ILM30211-C01EA 427 DKK
ILM30211-C01 ILM30211-C05 ILM30211-C1 VWRKILM30211-R10 VWRKILM30211-R25 VWRKS/ILM30211-R2
Anti-MS4A1 Mouse Monoclonal Antibody [clone: L26]
Antistoffer
Anti-CD20 (B-cell Pan) reacts with a membrane antigen that is present in B-cells. This antibody recognizes Reed-Sternberg cells, predominant in Hodgkin’s disease, in a minority of cases. Since no staining of histiocytes or plasma cells has been observed and CD20 has not been detected in T-cell malignancies, it is a very strong marker of B-cell lymphomas. Anti-CD20 does not cross-react with non-hematopoetic neoplasms.
  • Membranous visualization
  • Control: tonsil, lymph node
  • Dilution Range: 1:100 - 1:500
Pretreatment: Heat induced epitope retrieval in 10 mMMore Product Information