Anti-IGGL Mouse Monoclonal Antibody [clone: LAMB14]

Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ILM8283-C01 ILM8283-C05 ILM8283-C1 ILM8283-R10 ILM8283-R25
ILM8283-C01EA 611 DKK
ILM8283-C01 ILM8283-C05 ILM8283-C1 VWRKILM8283-R10 VWRKILM8283-R25
Anti-IGGL Mouse Monoclonal Antibody [clone: LAMB14]
Antistoffer
Anti-lambda detects surface immunoglobulin on normal and neoplastic B-cells. Its staining is seen in B-cell follicles of human lymphoid tissue. When dealing with B-cell neoplasms, the determination of light chain ratios remains helpful. Most B-cell lymphomas express either kappa or lambda light chains, whereas reactive proliferations display a mixture of kappa and lambda positive cells. If only a single light chain type is detected, a lymphoproliferative disorder is very likely.1-3

  • Visualization: cytoplasmic
  • Control: tonsil
  • Dilution Range: 1:100-1:500

References
1. Abbondanzo SL, et al. Ann Diagn Pathol. 1999; 3:318-27.
2. Kurtin PJ, et al. Am J Clin Pathol. 1999; 112:319-29.
3. Ashton-Key M, et al. Histopathology. 1996; 29:525-31.

Type: Primary
Antigen: IGGL
Clonality: Monoclonal
Clone: LAMB14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR