ESD safety ankle boots, lace-up, Trainers, Sander Pro
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
766771/38 766771/39 766771/40 766771/41 766771/42 766771/43 766771/44 766771/45 766771/46 766771/47
ELTN766771/38EA 679 DKK
ELTN766771/38 ELTN766771/39 ELTN766771/40 ELTN766771/41 ELTN766771/42 ELTN766771/43 ELTN766771/44 ELTN766771/45 ELTN766771/46 ELTN766771/47
ESD safety ankle boots, lace-up, Trainers, Sander Pro
Sko og støvler
100% metal- and leather-free, lightweight ESD safety trainer made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), a padded tongue plus a slip resistant, dual-density TPU/PU sole (Trainers).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length fleece inlay sole (ESD Pro), black
  • TPU scuff cap protects the leather upper
  • Metal-free puncture resistant midsole
  • Composite toe protection cap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/red
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.