ESD safety shoes, lace-up, Senex Pro
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
726771/38 726771/39 726771/40 726771/41 726771/42 726771/43 726771/44 726771/45 726771/46 726771/47
ELTN726771/38EA 656 DKK
ELTN726771/38 ELTN726771/39 ELTN726771/40 ELTN726771/41 ELTN726771/42 ELTN726771/43 ELTN726771/44 ELTN726771/45 ELTN726771/46 ELTN726771/47
ESD safety shoes, lace-up, Senex Pro
Sko og støvler
100% metal- and leather-free, sporty and lightweight ESD safety shoe made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), a padded tongue plus a slip resistant, dual-density TPU/PU sole (Trainers).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length fleece inlay sole (ESD Pro), black
  • TPU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Composite toe protection cap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/red
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC