Cooling circulation thermostats, PRO
Leverandør: LAUDA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L000021 L000022 L000023 L000024 L000102 L000104 L000122 L000124 L000182 L000184 L000202 L000204 L002494 L002495 L002502 L002503
461-0953EA 75990 DKK
461-0953 461-0954 461-0955 461-0956 LAUDL000102 LAUDL000104 LAUDL000122 LAUDL000124 LAUDL000182 LAUDL000184 LAUDL000202 LAUDL000204 461-1378 461-1379 461-1381 461-1382
Cooling circulation thermostats, PRO
Cirkulatorsystemer Lukkede cirkulatorsystemer Køletermostatbade
The PRO cooling circulation thermostats are ideal for external applications where rapid temperature changes are required. Their cooling output of 0,6 to 1,5 kW, combined with a very small filling volume enable these rapid temperature changes.
  • Touchscreen can be operated with gloves
  • Low filling volume combined with Varioflex pump ensures rapid temperature changes
  • Programmer with one program, 20 segments whereas 100 programs, with 50 segments for Command unit
  • Enables ramp function for easy computation
  • Hybrid cooling for cooling the refrigeration machine with ambient air or additional cooling water
  • USB and Ethernet interface equipped as standard
All devices use natural refrigerants and are fitted with a pressure-suction pump. It has detachable control panel for remote control. High contrast OLED display for units with Base control, whereas large 5,7" colour touchscreen (145 mm) with multi touch operation for units with Command Touch, with timer and count down display.
Control unit Base has menu navigation in plain text and operation via push buttons.
Control unit Command Touch has a weekly timer and count down function, ramp function, individual access rights for up to 20 users and data logging, export to a USB stick.
Max. flow rate/max. pressure:
Pump pressure/suction: 0,7/0,4 bar
Pump flow pressure/suction: 22/20 L/min
Leveringsinformation: Supplied with 2 hose olives (13 mm) for pump connection and 2 hose olives (10 mm) with cap nut G3/8 for cooling water connection.