ESD safety shoes, lace-up, Safety-Grip, Renzo XW Low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
725861/44 725861/45 725861/46 725861/47 725861/48 725861/49 725861/50 725861/36 725861/37 725861/38 725861/39 725861/40 725861/41 725861/42 725861/43
ELTN725861/44EA 485 DKK
ELTN725861/44 ELTN725861/45 ELTN725861/46 ELTN725861/47 ELTN725861/48 ELTN725861/49 ELTN725861/50 ELTN725861/36 ELTN725861/37 ELTN725861/38 ELTN725861/39 ELTN725861/40 ELTN725861/41 ELTN725861/42 ELTN725861/43
ESD safety shoes, lace-up, Safety-Grip, Renzo XW Low
Sko og støvler
Wide, durable ESD safety shoes made from robust, breathable cowhide leather, reflective materials, a padded bellows tongue and collar, plus a dual-density PU/PU sole (Saftey-Grip) (3,5 mm) with exceptional non slip and good self-cleaning properties.
  • Climate-regulating, breathable textile lining
  • Full length fleece inlay sole (ESD Pro), black
  • Sole matches the movement of the foot, for more stability and less fatigue
  • PU scuff cap protects the leather upper
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
  • Extra wide version (XW)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.