ESD safety shoes, lace-up, Safety-Grip, Renzo Low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
725841/36 725841/37 725841/38 725841/39 725841/40 725841/41 725841/42 725841/43 725841/44 725841/45 725841/46 725841/47 725841/48 725841/49 725841/50
114-7679EA 485 DKK
114-7679 114-7680 114-7681 114-7682 114-7683 114-7684 114-7685 114-7686 114-7687 114-7688 114-7689 114-7690 114-7691 114-7692 114-7693
ESD safety shoes, lace-up, Safety-Grip, Renzo Low
Sko og støvler
Durable ESD safety shoes made from robust, breathable cowhide leather, reflective materials, a padded bellows tongue and collar, plus a dual-density PU/PU tread sole (Safety-Grip) (3,5 mm) with exceptional non slip and good self-cleaning properties.
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length fleece inlay sole (ESD Pro), black
  • Sole matches the movement of the foot, for more stability and less fatigue
  • PU scuff cap protects the leather upper
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.