ESD safety shoes, lace-up, Fun, Maverick Blue Low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
723351/38 723351/39 723351/40 723351/41 723351/42 723351/43 723351/44 723351/45 723351/46 723351/47 723351/48
ELTN723351/38EA 611 DKK
ELTN723351/38 ELTN723351/39 ELTN723351/40 ELTN723351/41 ELTN723351/42 ELTN723351/43 ELTN723351/44 ELTN723351/45 ELTN723351/46 ELTN723351/47 ELTN723351/48
ESD safety shoes, lace-up, Fun, Maverick Blue Low
Sko og støvler
Sporty, lightweight, leather-free ESD safety shoe made from microfibre and mesh inserts, a padded tongue plus a slip resistant, dual-density PU/PU sole (Fun).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), black
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Blue/white
Certificeringer: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC