VWR® Traceable® excursion trac, Data Logging ULT Freezer Thermometers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-2440EA 2730 DKK
620-2440 620-2441 620-2444 620-2445
VWR® Traceable® excursion trac, Data Logging ULT Freezer Thermometers
Termometre Digital termometre
Data logging thermometers with user-defined timing intervals. Ideal for monitoring ultra-low temperature freezers.
  • Simultaneously display, minimum, maximum and current temperatures, °F/°C switchable
  • Rolling memory structure, maintains the most recent 525 000 temperature observations and 10 unique alarm events
  • Recorded data (CSV file) may be transferred from thermometer to PC or Mac using a USB flash drive - no additional software is needed
  • Memory can be cleared on unit after downloading data to USB stick
  • Smart alarm features a visual/audible alarm that continues to alarm even if the unit returns to non alarm conditions
  • Status indicators - low battery, memory full, USB data transfer and active alarm state
  • Includes 24,5 cm PTFE cable
  • Hassle-free retrieval of data - unit can remain in use while downloading and analysing data
Certificeringer: Thermometer is calibrated at multiple points (−80; −40; 0 and +100 °C) and comes with an individually serialised and certified Traceable® calibration certificate from an ISO/IEC 17025 calibration laboratory accredited by A2LA as standard.
Leveringsinformation: Supplied with stand, batteries, VELCRO®, magnetic strips, wall mount and Traceable® certificate.