Jars, wide mouth, depyrogenated glass
Leverandør: EP SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
130-04C/PF 130-04A/PF 132-16C/PF 130-04C/TL/PF 131-08A/PF 131-08C/PF 130-02C/PF C20-02A/PF
EPSC130-04C/PFEA 3390 DKK
EPSC130-04C/PF EPSC130-04A/PF EPSC132-16C/PF EPSC130-04C/TL/PF EPSC131-08A/PF EPSC131-08C/PF EPSC130-02C/PF EPSCC20-02A/PF
Jars, wide mouth, depyrogenated glass
Dåser Dåser, allround
Clear or amber glass with closure.
  • Packaging and storing articles that will be terminally sterilized
  • Storage of laboratory reagents and medias
  • Sample storage and water sampling
  • Meet endotoxin levels of less than 0,06 EU/ml
Leveringsinformation: It is supplied with certificate of analysis.