Accessories for ULT freezer TSX400V
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FCR567TSX SS34567 FCR400TSX FCRP400TSX SK400TSX
HERAFCR567TSXEA 16510 DKK
HERAFCR567TSX REVCSS34567 471-1171 471-1172 471-1173
Accessories for ULT freezer TSX400V
Frysere Ultra lav temperatur frysere
Record seven days of temperature performance at temperatures to −150 °C using Thermo Scientific™ chart recorders for ultra-low temperature freezers. Recorders use 150 mm chart paper and are available as factory or field installed.
  • Seven-day recorder
  • Field or factory installed
  • Field-installed units must be installed by a qualified professional