3-(2-Bromoethyl)-1H-indole 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-(2-Bromoethyl)indole

426910010 426910050
ACRO426910010EA 379 DKK
ACRO426910010 ACRO426910050
3-(2-Bromoethyl)-1H-indole 95%
3-(2-Bromoethyl)-1H-indole

Appearance: Off-white to light brown Crystalline powder

Formel: C₁₀H₁₀BrN
Smeltepunkt: 97…99 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00130155
CAS-nummer: 3389-21-7
UN nummer: 3261
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance Off-white to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 95 to 101 °C
GC ≥94.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR