1-(2-Bromethyl)naphthalen 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

399560010 399560050
ACRO399560010EA 800 DKK
ACRO399560010 ACRO399560050
1-(2-Bromethyl)naphthalen 97%
1-(2-Bromethyl)naphthalen
Formel: C₁₂H₁₁Br
Flammepunkt: 110 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00037737
CAS-nummer: 13686-49-2

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6380 to 1.6400 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR