4-Iodphenylhydrazin 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
399050010
399050050
ACRO399050010EA
418
DKK
ACRO399050010
ACRO399050050
4-Iodphenylhydrazin 95%
4-Iodphenylhydrazin
Formel:
C₆H₇IN₂ Smeltepunkt: 101…102 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00052688 CAS-nummer: 13116-27-3 UN nummer: 2811 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | White to orange to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 99 to 106 °C |
NMR | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...