1-Isopropylpiperazin 98+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

384810010 384810050
ACRO384810010EA 273 DKK
ACRO384810010 ACRO384810050
1-Isopropylpiperazin 98+%
1-Isopropylpiperazin
Formel: C₇H₁₆N₂
Flammepunkt: 54 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00167971
CAS-nummer: 4318-42-7
UN nummer: 2733
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4720 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR