Furan 99+% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

181110010 181112500 181110025
ACRO181110010EA 727 DKK
ACRO181110010 ACRO181112500 ACRO181110025
Furan 99+% stabilised
Furan
Formel: C₄H₄O
Kogepunkt: 31,3 °C
Massefylde: 0,936 g/cm³
Flammepunkt: –35 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003222
CAS-nummer: 110-00-9
UN nummer: 2389
ADR: 3,I

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4197 to 1.4217 (20°C, 589 nm)
Water ≤0.2 %
GC ≤0.05 % (Furfural)
Stabilizer 0.025 to 0.04 % BHT
Peroxides ≤0.015 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR