Ammonium bis(oxalato)oxotitanate(IV) hydrate 98%, pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

212730100 212731000
ACRO212730100EA 710 DKK
ACRO212730100 ACRO212731000
Ammonium bis(oxalato)oxotitanate(IV) hydrate 98%, pure
Ammonium bis(oxalato)oxotitanate(IV) hydrate

CAS No.: 10580-03-7

Formel: (NH₄)₂TiO(C₂O₄)₂·xH₂O (x≈1)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149620
CAS-nummer: 10580-03-7

Bestil nu

Specifikationer Testresultater

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Titration with KMnO4 ≥97.5 % (Calculated on oxalate)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR