2-(Aminomethyl)phenol 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Hydroxybenzylamine

298890010 298890050
ACRO298890010EA 1780 DKK
ACRO298890010 ACRO298890050
2-(Aminomethyl)phenol 98%
2-(Aminomethyl)phenol

CAS No.: 932-30-9

Formel: C₇H₉NO
Smeltepunkt: 127…131 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00870498
CAS-nummer: 932-30-9

Bestil nu

Specifikationer Testresultater

Appearance Beige Powder
Infrared spectrum Conforms
Melting point 127 to 131 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR