Aluminiumoxid acidic Brockmann Grade I 40-300 µm, 60A til kromotografi
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
365750010 365750025 365750100
ACRO365750010EA 991 DKK
ACRO365750010 ACRO365750025 ACRO365750100
Aluminiumoxid acidic Brockmann Grade I 40-300 µm, 60A til kromotografi
Aluminiumoxid acidic
Formel: Al₂O₃
Kogepunkt: 2980 °C (760 mmHg)
Smeltepunkt: 2030 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003424
CAS-nummer: 1344-28-1
Bestil nu
Specifikationer Testresultater
Appearance White to off-white Powder
Particle size ≤5 % (<40 µm)
Specific surface area 185 to 225 m²/g
pH 4.0 to 5.0 (10% aqueous suspension)
Sodium oxide (Na2O) ≤0.4 %
Silicon dioxide (SiO2) ≤0.03 %
Iron(III)oxide (Fe2O3) ≤0.03 %