Blytetraacetat 95% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Lead(IV) tetraacetate, Triacetyloxyplumbyl acetate, Acetic acid lead (IV) salt, Lead (IV) acetate, LTA

180621000 180625000 180620025
ACRO180621000EA 465 DKK
ACRO180621000 ACRO180625000 ACRO180620025
Blytetraacetat 95% stabilised
Blytetraacetat
Formel: (H₃CCOO)₄Pb
Molekylvægt: 443.38 g/mol
Smeltepunkt: 175…180 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00008693
CAS-nummer: 546-67-8
UN nummer: 1616
ADR: 6.1 III
Merck Index: 13,05442

Bestil nu

Specifikationer Testresultater

Appearance White to pink-orange Crystalline powder
Titration Iodometric ≥94 %
Stabilizer ca. 4 to 5 % Acetic acid

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR