Polysulphone resin, M.W. ≈. M.W. 60,000, piller

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

178910050 178912500
ACRO178910050EA 380 DKK
ACRO178910050 ACRO178912500
Polysulphone resin, M.W. ≈. M.W. 60,000, piller
Polysulphone resin

Appearance: Amber to beige Pellets

Massefylde: 1,24 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134387
CAS-nummer: 25135-51-7

Bestil nu

Specifikationer Testresultater

Appearance Amber to beige Pellets
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR