Brucine dihydrate 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

163500500 163501000
ACRO163500500EA 577 DKK
ACRO163500500 ACRO163501000
Brucine dihydrate 99%
Brucine dihydrate

CAS No.: 145428-94-0

Formel: C₂₃H₂₆N₂O₄·2H₂O
Molekylvægt: 430.50 g/mol (394.47 anhy)
Smeltepunkt: 174…179 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149384
CAS-nummer: 145428-94-0
UN nummer: 1570
ADR: 6.1 I

Bestil nu

Specifikationer Testresultater

Appearance White to light beige Powder or crystals
Infrared spectrum Conforms
Melting point 174 to 179 °C
Loss on drying 6.0 % to 9.5 % (105 °C, 3 hrs)
Specific optical rotation -75° to -80° (20 °C, 589 nm) (c=1, EtOH)
Titration with HClO4 ≥98.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR