Cholesterol pelargonate 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Cholesterol Nonanoate, Nonanoic Acid Cholesterol Ester, Pelargonic Acid Cholesterol Ester

110270250 110271000
ACRO110270250EA 584 DKK
ACRO110270250 ACRO110271000
Cholesterol pelargonate 99+%
Cholesterol pelargonate

Appearance: White to off-white Powder

Formel: C₃₆H₆₂O₂
Kogepunkt: 360 °C (760 mmHg)
Smeltepunkt: 74…77 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003643
CAS-nummer: 1182-66-7

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
TLC ≥99.0 %
Appearance of solution (10% soln. in hexane) clear, colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR