L-Azetidine-2-carboxylic acid 99+%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
105142500
105140010
105140050
105141000
ACRO105142500EA
718
DKK
ACRO105142500
ACRO105140010
ACRO105140050
ACRO105141000
L-Azetidine-2-carboxylic acid 99+%
L-Azetidine-2-carboxylic acid
CAS No.: 2133-34-8
Formel:
C₄H₇NO₂ Smeltepunkt: 206…207 °C (dec.) Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00005166 CAS-nummer: 2133-34-8 |
Specifikationer Testresultater
Appearance | White to light beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
TLC | ≥99.0 % |
Specific optical rotation | -123° to -127° (25°C, 589 nm) (c=1, water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...