Aminoguanidine nitrate 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
103760050
103761000
103760250
ACRO103760050EA
160
DKK
ACRO103760050
ACRO103761000
ACRO103760250
Aminoguanidine nitrate 99%
Aminoguanidine nitrate
Appearance: White Crystalline powder
Formel:
CH₇N₅O₃ Molekylvægt: 137.099 g/mol Smeltepunkt: 145…147 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00013174 CAS-nummer: 10308-82-4 UN nummer: 1479 ADR: 5.1 II |
Specifikationer Testresultater
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 143 to 147 °C |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...