4-Amino-6-chloro-2-(methylthio)pyrimidine 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Amino-6-chloro-2-methylmercaptopyrimidine
103390050
103390250
103390010
ACRO103390050EA
918
DKK
ACRO103390050
ACRO103390250
ACRO103390010
4-Amino-6-chloro-2-(methylthio)pyrimidine 97%
4-Amino-6-chloro-2-(methylthio)pyrimidine
CAS No.: 1005-38-5
Formel:
C₅H₆ClN₃S Smeltepunkt: 130…134 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006088 CAS-nummer: 1005-38-5 |
Specifikationer Testresultater
Appearance | Off-white to yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 130 to 134 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...