Rhodamine 123 99+%, pure
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 3,6-Diamino-9-(2-(methoxycarbonyl)phenyl xanthylium chloride, 3,6-Diamino-9-(2-(methoxycarbonyl)phenyl)xanthylium chloride, 2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid methyl ester
419091000 419095000
ACRO419091000EA 2160 DKK
ACRO419091000 ACRO419095000
Rhodamine 123 99+%, pure
Rhodamine 123
Appearance: Red-brown to brown Powder
Formel: C₂₁H₁₇ClN₂O₃
Molekylvægt: 380.82 g/mol
Smeltepunkt: 235 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012664
CAS-nummer: 62669-70-9
Bestil nu
Specifikationer Testresultater
Appearance Red-brown to brown Powder
Infrared spectrum Conforms
UV spectrophotometry ≥99.0 %