Naphthenic acid pract.

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Naphthenic acids

415281000 415285000 415280025
ACRO415281000EA 161 DKK
ACRO415281000 ACRO415285000 ACRO415280025
Naphthenic acid pract.
Naphthenic acid

CAS No.: 1338-24-5

Kogepunkt: 140…200 °C (760 mmHg)
Massefylde: 0.91 g/cm³
Flammepunkt: 149 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00147696
CAS-nummer: 1338-24-5

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Density (20°C) 910 to 940 g/l
Viscosity ≤5 St (at 25°C)
Acid value 230 to 260 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR