N-tert-Butylacrylamide 97%, ren
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
403451000
403455000
ACRO403451000EA
859
DKK
ACRO403451000
ACRO403455000
N-tert-Butylacrylamide 97%, ren
N-tert-Butylacrylamide
Appearance: White to off-white Powder and/or chunks
Formel:
C₇H₁₃NO Smeltepunkt: 128…130 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00026271 CAS-nummer: 107-58-4 |
Specifikationer Testresultater
Appearance | White to off-white Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 126 to 131 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...