8-hydroxyquinoline-2-carboxylic acid 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

347620010 347620050
ACRO347620010EA 832 DKK
ACRO347620010 ACRO347620050
8-hydroxyquinoline-2-carboxylic acid 98%
8-hydroxyquinoline-2-carboxylic acid

Appearance: Yellow Crystalline powder

Formel: C₁₀H₇NO₃
Smeltepunkt: 213…214 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00168972
CAS-nummer: 1571-30-8

Bestil nu

Specifikationer Testresultater

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 219 to 223 °C
Titration with TBAH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR