Tetra-tert-butyl orthotitanate 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Titanium(IV) tert-butoxide, Titanic acid tetra-tert-butyl ester, Titanium(IV) t-butoxide, Titanium tetra-tert-butylate, Orthotitanic acid tetra-tert-butyl ester, tert-Butyl titanate(IV)
344000050
344000250
ACRO344000050EA
685
DKK
ACRO344000050
ACRO344000250
Tetra-tert-butyl orthotitanate 98%
Tetra-tert-butyl orthotitanate
Appearance: Clear colorless to light yellow Liquid
Formel:
C₁₆H₃₆O₄Ti Kogepunkt: 160…165 °C (15 mmHg) Massefylde: 0,8893 g/cm³ Flammepunkt: 47 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00040554 CAS-nummer: 3087-39-6 UN nummer: 1993 ADR: 3,III |
Specifikationer Testresultater
Appearance | Clear colorless to light yellow Liquid |
Assay | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...