Paclitaxel 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

328420010 328420050 328420250
ACRO328420010EA 539 DKK
ACRO328420010 ACRO328420050 ACRO328420250
Paclitaxel 99+%
Paclitaxel

CAS No.: 33069-62-4

Formel: C₄₇H₅₁NO₁₄
Smeltepunkt: 213…216 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00869953
CAS-nummer: 33069-62-4

Bestil nu

Specifikationer Testresultater

Appearance White Powder
Infrared spectrum Conforms
Melting point 203 to 223 °C
HPLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR