2-Cyanothioacetamide 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

305680050 305680250
ACRO305680050EA 435 DKK
ACRO305680050 ACRO305680250
2-Cyanothioacetamide 97%
2-Cyanothioacetamide

Appearance: Beige to brown Powder or needles

Formel: C₃H₄N₂S
Molekylvægt: 100.139 g/mol
Smeltepunkt: 113…119 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010025
CAS-nummer: 7357-70-2
UN nummer: 2811
ADR: 6.1 III

Bestil nu

Specifikationer Testresultater

Appearance Beige to brown Powder or needles
Infrared spectrum Conforms
Melting point 113 to 119 °C
Titration with TBAH 96.0 to 104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR