(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Cbz-Phe-OH, N-Cbz-L-phenylalanine, N-Carbobenzyloxy-L-phenylalanine, Z-Phe-OH

275960050 275960250
ACRO275960050EA 249 DKK
ACRO275960050 ACRO275960250
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid 99+%
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid

Appearance: White to off-white Powder

Formel: C₁₇H₁₇NO₄
Smeltepunkt: 85…88 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00020418
CAS-nummer: 1161-13-3

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 84 to 88 °C
Water ≤0.2 % (K.F.)
HPLC ≥98.0 %
Specific optical rotation +4.5° to +5.5° (20 °C, 589 nm) (c=5, CH3COOH)
Titration with NaOH 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR