Boc-D-Alanine 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-Boc-D-alanine, Boc-D-Ala-OH

275720050 275720250
ACRO275720050EA 373 DKK
ACRO275720050 ACRO275720250
Boc-D-Alanine 99+%
Boc-D-Alanine

CAS No.: 7764-95-6

Formel: C₈H₁₅NO₄
Smeltepunkt: 81…84 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00063123
CAS-nummer: 7764-95-6

Bestil nu

Specifikationer Testresultater

Appearance White Powder
Infrared spectrum Conforms
Melting point 81 to 84 °C
HPLC ≥99.0 % (ELSD)
Specific optical rotation +25° to +27° (20 °C, 589 nm) (c=2, CH3COOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR