D-Tagatose 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

257890010 257892500 257890050
ACRO257890010EA 1130 DKK
ACRO257890010 ACRO257892500 ACRO257890050
D-Tagatose 99+%
D-Tagatose

Appearance: White to off-white Crystalline powder

Formel: C₆H₁₂O₆
Smeltepunkt: 129…133 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134449
CAS-nummer: 87-81-0

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1.0 %
Sulfated ash ≤0.1 %
Solubility (5 % in water) Clear colorless to almost colorless
HPLC ≥99.0 % (ELSD)
Specific optical rotation -5° to -7° (20°C, 589 nm) (c=2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR