Sesame oil

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

241000010 241002500
ACRO241000010EA 904 DKK
ACRO241000010 ACRO241002500
Sesame oil
Sesame oil

CAS No.: 8008-74-0

Smeltepunkt: –5 °C
Massefylde: 0.919 g/cm³
Flammepunkt: 255 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00132243
CAS-nummer: 8008-74-0

Bestil nu

Specifikationer Testresultater

Appearance Clear yellow Oily liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4760 (20°C, 589 nm)
Density (20°C) 0.915 to 0.923
Acid value ≤0.6 mg KOH/g
Peroxide value ≤10 meq/kg
Unsaponifiable matter ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR