Anti-IgG Donkey Polyclonal Antibody (CF™ 680)

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

20417-50UL 20417-250UL
20417-50ULEA 1130 DKK
20417-50UL 20417-250UL
Anti-IgG Donkey Polyclonal Antibody (CF™ 680)
Antistoffer
This product is prepared by labeling highly cross-adsorbed donkey anti-rat IgG (H+L) with our near-IR fluorescent dye CF™680. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, horse, human, mouse, rabbit, and sheep serum proteins. Far red/near-infrared CF™680 (Ex/Em 681/698 nm) is brighter and produces antibody conjugates with the best signal-to-noise ratio compared to other spectrally similar dyes.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF680
Epitope:
Host: Donkey
Isotype:
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR